Learn More
Medchemexpress LLC Sant-1 | 304909-07-7 | MFCD01827306 | 99.8% | 373.49 g/mol | C23H27N5 | 100 MG
SDP
Catalog No. 5000215779
Encompass_Preferred
SANT-1 is a small-molecule antagonist of the Smoothened (Smo) receptor that inhibits Hedgehog signaling. It demonstrates low-nanomolar activity in cell-based assays and is used to probe Smo-mediated signaling in cellular models.
  • Potent Smoothened (Smo) antagonist with low-nanomolar IC50s (20 nM and 30 nM).
  • Reduces proliferation of medulloblastoma cells at 1 μM.
  • High purity (~99.8%) suitable for research applications.
  • Soluble in DMSO (25 mg/mL) with documented in vivo formulation protocols.
  • Powder stable at -20°C for extended storage.
  • Available in multiple package sizes up to 500 mg.
Catalog No. 50-002-15779 Supplier Medchemexpress LLC Supplier No. HY100224100MG
SANT-1 is a small-molecule antagonist of the Smoothened (Smo) receptor that inhibits Hedgehog signaling. It demonstrates low-nanomolar activity in cell-based assays and is used to probe Smo-mediated signaling in cellular models.
  • Potent Smoothened (Smo) antagonist with low-nanomolar IC50s (20 nM and 30 nM).
  • Reduces proliferation of medulloblastoma cells at 1 μM.
  • High purity (~99.8%) suitable for research applications.
  • Soluble in DMSO (25 mg/mL) with documented in vivo formulation protocols.
  • Powder stable at -20°C for extended storage.
  • Available in multiple package sizes up to 500 mg.