Learn More
AdipoGen SANT-2 (1 mg)
SDP
Supplier:  AdipoGen AGCR13514M001
Encompass_Preferred
Chemical. CAS: 329196-48-7. Formula: C26H26ClN3O4. MW: 480. Sonic hedgehog (Shh) signaling pathway modulator. Cell permeable potent smoothened (Smo) antagonist. Inhibits wt and oncogenic Smo. SAG activity (Prod. Nr. AG-CR1-3506) inhibitor. Gli1 reporter gene expression inhibitor. Suppresses the Shh-dependent translocation of Smo to the primary cilium. Potential tumor growth blocker.
Catalog No. 50-205-3893