Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eMolecules​ Santa Cruz Biotechnology 2-(1H-Imidazol-1-yl)ethanamine, C5H9N3, 111.15 g/mol, 1 ea

Catalog No. NC3940660 Shop All eMolecules Inc. Products
Encompass_Preferred

2-(1H-Imidazol-1-yl)ethanamine, also known as 2-imidazol-1-yl-ethylamine, is a chemical compound with the molecular formula C5H9N3 and a molecular weight of 111.15. This product is intended strictly for research purposes.

  • Molecular formula of C5H9N3
  • Molecular weight of 111.15
  • For research use only
  • Not intended for diagnostic or therapeutic use

Catalog No. NC3940660 Supplier eMolecules​ Supplier No. 937463098
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-(1H-Imidazol-1-yl)ethanamine, also known as 2-imidazol-1-yl-ethylamine, is a chemical compound with the molecular formula C5H9N3 and a molecular weight of 111.15. This product is intended strictly for research purposes.

  • Molecular formula of C5H9N3
  • Molecular weight of 111.15
  • For research use only
  • Not intended for diagnostic or therapeutic use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.