Learn More
Selleck Chemical LLC Santacruzamate A
SDP
Supplier:  Selleck Chemical LLC S759550MG
Encompass_Preferred
Santacruzamate A (CAY10683) is a potent and selective HDAC inhibitor with IC50 of 119 pM for HDAC2 3600-fold selectivity over other HDACs
Catalog No. 50-000-04827