Learn More
Selleck Chemical LLC Santacruzamate A
SDP
Catalog No. 5000004828
Encompass_Preferred
Santacruzamate A (CAY10683) is a potent and selective HDAC inhibitor with IC50 of 119 pM for HDAC2 3600-fold selectivity over other HDACs
Catalog No. 50-000-04828 Supplier Selleck Chemical LLC Supplier No. S759510MM/1ML
Santacruzamate A (CAY10683) is a potent and selective HDAC inhibitor with IC50 of 119 pM for HDAC2 3600-fold selectivity over other HDACs