Learn More
Apexbio Technology LLC Santacruzamate A (CAY10683) 1477949-42-0 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B47945.1
Encompass_Preferred
Santacruzamate A (CAY10683 CAS 1477949-42-0) is a selective histone deacetylase (HDAC) inhibitor originally identified in the marine cyanobacterium cf Symploca sp Its mechanism of action involves targeting HDAC enzymes particularly demonstrating high selectivity towards HDAC2 (IC 0 112 nM) when compared with HDAC6 (IC 433 nM) and minimal potency against HDAC4 (IC 1 M) In cellular assays Santacruzamate A reduces growth of tumor cell lines such as HCT-116 colon cancer cells (GI 28 M) and HuT-78 cutaneous T-cell lymphoma cells (GI 1 3 M) It is valuable in epigenetics and oncology research for selectively modulating HDAC2 activity
Catalog No. 50-000-62717