Learn More
Medchemexpress LLC HY-N0931 5mg Medchemexpress, Santacruzamate A CAS:1477949-42-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN09315MG
Encompass_Preferred
Medchemexpress, HY-N0931 5mg Santacruzamate A CAS:1477949-42-0 Santacruzamate A (CAY-10683) is a potent and selective HDAC2 inhibitor with an IC50 of 119 pM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1416