Learn More
Cole-Parmer Fitting, 304 Stainless Steel, Sanitary Clamp, 1/2" to 3/4"
Catalog No. 502557746 Shop All Cole-Parmer Products
Encompass_Preferred
Use the sanitary clamps with sanitary fittings to create a leak-proof connection. These easy-to-use clamps are made from 304 stainless steel to provides good chemical resistance.
Catalog No. 50-255-7746 Supplier Cole-Parmer Supplier No. 5011842
Use the sanitary clamps with sanitary fittings to create a leak-proof connection. These easy-to-use clamps are made from 304 stainless steel to provides good chemical resistance.