Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Santonin, MP Biomedicals™

Catalog No. MP215658210 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215658210 10 g
MP215658250 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP215658210 Supplier MP Biomedicals, Inc Supplier No. 0215658210

Santonin, MP Biomedicals is a chiral building block used for the synthesis of various sesquiterpenoides.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 481-06-1
Molecular Formula C15H18O3
Molecular Weight (g/mol) 246.31
MDL Number MFCD00135865
InChI Key XJHDMGJURBVLLE-BOCCBSBMSA-N
Synonym santonin, alpha-santonin, --alpha-santonin, santoninic anhydride, --santonin, unii-1vl8j38ero, semenen, --santonine, 1vl8j38ero, santoninum
PubChem CID 221071
ChEBI CHEBI:16363
IUPAC Name (3S,3aS,5aS,9bS)-3,5a,9-trimethyl-2H,3H,3aH,4H,5H,5aH,8H,9bH-naphtho[1,2-b]furan-2,8-dione
SMILES C[C@H]1[C@@H]2CC[C@@]3(C)C=CC(=O)C(C)=C3[C@H]2OC1=O

Specifications

Melting Point 172°C to 173°C (literature)
Quantity 10 g
Solubility Information Soluble in cold water, boiling water, alcohol cold/boiling, cold/boling ether, chloroform (50mg/mL).
Optical Rotation [α]20/D −170 ± 5° (c=1% in ethanol) (Lit.)
Formula Weight 246.31
Percent Purity ≥98%
Physical Form Crystalline Powder
Recommended Storage 4°C, desiccate, protect from light
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.