Learn More
Electron Microscopy Sciences Santovac 5 for Ultra High Vacuum Range 100 ML
SDP
Supplier:  Electron Microscopy Sciences 60730
Encompass_Preferred
A Polymethyl Ether, designed for an ultra high vacuum range: 10-10 torr. Ultra low vapor pressure results in low ultimate pressures; there is no need for a cold trap and it is suitable for diffusion pumps. Santovac 5 may discolor with use but it will maintain its physical properties.
Catalog No. 50-996-274