Learn More
Electron Microscopy Sciences Santovac 5 for Ultra High Vacuum Range 500 ML
SDP
Catalog No. 50996275
Encompass_Preferred
A Polymethyl Ether, designed for an ultra high vacuum range: 10-10 torr. Ultra low vapor pressure results in low ultimate pressures; there is no need for a cold trap and it is suitable for diffusion pumps. Santovac 5 may discolor with use but it will maintain its physical properties.
Catalog No. 50-996-275 Supplier Electron Microscopy Sciences Supplier No. 60732
A Polymethyl Ether, designed for an ultra high vacuum range: 10-10 torr. Ultra low vapor pressure results in low ultimate pressures; there is no need for a cold trap and it is suitable for diffusion pumps. Santovac 5 may discolor with use but it will maintain its physical properties.