Learn More
Aviva Systems Biology Corp SAP18 Antibody - N-terminal region (ARP38690_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38690_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SAP18. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SAP18. Alternate names: SAP18P, 2HOR0202ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56756