Learn More
Aviva Systems Biology Corp SAP30 Antibody - N-terminal region (ARP38392_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38392_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SAP30. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SAP30. Alternate names: ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56635