Learn More
Aviva Systems Biology Corp SAP30BP Antibody - N-terminal region (ARP39013_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39013_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SAP30BP. It was validated on Western Blot. . Alternate names: DKFZp586L2022, HCNGP, HTRG, HTRPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-284-773