Learn More
Aviva Systems Biology Corp SAP30BP Antibody - C-terminal region (ARP34303_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34303_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SAP30BP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human SAP30BP. Alternate names: HTRG, HTRP, HCNGPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55333