Learn More
Enzo Life Sciences SAP 97 monoclonal antibody (RPI 197.4) (200 µg)
SDP
Supplier:  Enzo Life Sciences ADIVAMPS005F
Encompass_Preferred
Clone: RPI 197.4. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant rat SAP97 (N-terminal fragment). UniProt: Q62696. Source: Purified from ascites. Species Reactivity: Human, Mouse, Rat, Dog. Applications: ICC, IHC, IP, WB, Electron microscopy. Recommended dilutions: Western Blot (1:1,000, ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-667-834