Learn More
Enzo Life Sciences SAP97 polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences ALX210280R100
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to aa 115-133 (V115LPSERISPQVPNEVLGPE133) of rat SAP97 (synapse-associated protein 97). Species reactivity: Mouse, Rat. Applications: ICC, IHC (FS), IP, WB. Recommended Dilutions: Immunohistochemistry (frozen sections, 1:1,000) Western Blot (1:1,000) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Formulation: Liquid. In PBS containing 0.05% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-8103