Learn More
Cayman Chemical Sapacitabine, Purity: ≥95%, CAS: 151823-14-2, 10 mg
SDP
Catalog No. 502940932
Encompass_Preferred
A prodrug form of CNDAC; reduces tumor volume in an MV4-11 mouse xenograft model at 5 and 15 mg/kg alone or in combination with SAHA
Catalog No. 50-294-0932 Supplier Cayman Chemical Supplier No. 2732110 MG
A prodrug form of CNDAC; reduces tumor volume in an MV4-11 mouse xenograft model at 5 and 15 mg/kg alone or in combination with SAHA