Learn More
Medchemexpress LLC HY-13328 10mg Medchemexpress, Sapanisertib CAS:1224844-38-5 Purity:>98%
SDP
Catalog No. 502028984
Encompass_Preferred
Medchemexpress, HY-13328 10mg Sapanisertib CAS:1224844-38-5 Sapanisertib (INK-128; MLN0128; TAK-228) is an orally available, ATP-dependent mTOR1/2 inhibitor with an IC50 of 1 nM for mTOR kinase. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8984 Supplier Medchemexpress LLC Supplier No. HY1332810MG
Medchemexpress, HY-13328 10mg Sapanisertib CAS:1224844-38-5 Sapanisertib (INK-128; MLN0128; TAK-228) is an orally available, ATP-dependent mTOR1/2 inhibitor with an IC50 of 1 nM for mTOR kinase. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.