Learn More
New England Biolabs, Inc. SapI – 1250 units
SDP
Catalog No. 50812260
Encompass
A restriction endonuclease that recognizes the sequence GCTCTTCN^NNN_.
Catalog No. 50-812-260 Supplier New England Biolabs, Inc. Supplier No. R0569L
A restriction endonuclease that recognizes the sequence GCTCTTCN^NNN_.