Learn More
Medchemexpress LLC Sapintoxin D | 80998-07-8 | 99.9% | 10 MG
SDP
Catalog No. 5000341346
Encompass_Preferred
Sapintoxin D is a fluorescent phorbol ester used to study the activation of protein kinase C (PKC). It functions as a calcium-dependent PKC activator, making it suitable for various research applications in cell biology and biochemistry.
  • Fluorescent phorbol ester
  • Activates protein kinase C (PKC)
  • Calcium-dependent mechanism
  • Appears as a white to off-white solid
  • High purity of 99.91%
  • Soluble in DMSO for in vitro studies
Catalog No. 50-003-41346 Supplier Medchemexpress LLC Supplier No. HY12933010MG
Sapintoxin D is a fluorescent phorbol ester used to study the activation of protein kinase C (PKC). It functions as a calcium-dependent PKC activator, making it suitable for various research applications in cell biology and biochemistry.
  • Fluorescent phorbol ester
  • Activates protein kinase C (PKC)
  • Calcium-dependent mechanism
  • Appears as a white to off-white solid
  • High purity of 99.91%
  • Soluble in DMSO for in vitro studies