Learn More
Medchemexpress LLC Sapintoxin D | 80998-07-8 | 99.9% | 539.62 g/mol | C30H37NO8 | 5 MG
SDP
Catalog No. 5000262190
Encompass_Preferred
Sapintoxin D is a fluorescent phorbol ester used as a calcium-dependent activator of protein kinase C (PKC) for biochemical and cell-based research. It enables study of PKC activation, membrane translocation, and downstream phosphorylation events in signaling pathways, and is provided as a high-purity research reagent.
  • Fluorescent phorbol ester useful for visualization in assays
  • Calcium-dependent activation of protein kinase C
  • High purity suitable for research applications
  • Molecular formula C30H37NO8 and molecular weight 539.62 g/mol
  • Available in small milligram quantities for assay flexibility
  • Applicable to biochemical and cell-based PKC signaling studies
Catalog No. 50-002-62190 Supplier Medchemexpress LLC Supplier No. HY1293305MG
Sapintoxin D is a fluorescent phorbol ester used as a calcium-dependent activator of protein kinase C (PKC) for biochemical and cell-based research. It enables study of PKC activation, membrane translocation, and downstream phosphorylation events in signaling pathways, and is provided as a high-purity research reagent.
  • Fluorescent phorbol ester useful for visualization in assays
  • Calcium-dependent activation of protein kinase C
  • High purity suitable for research applications
  • Molecular formula C30H37NO8 and molecular weight 539.62 g/mol
  • Available in small milligram quantities for assay flexibility
  • Applicable to biochemical and cell-based PKC signaling studies