Learn More
Medchemexpress LLC HY-13050 10mg Medchemexpress, Sapitinib CAS:848942-61-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1305010MG
Encompass_Preferred
Medchemexpress, HY-13050 10mg Sapitinib CAS:848942-61-0 Sapitinib (AZD-8931) is a reversible, ATP competitive EGFR inhibitor of with IC50s of 4, 3 and 4 nM for EGFR, ErbB2 and ErbB3 in cells, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8954