Learn More
Medchemexpress LLC Sapitinib | 848942-61-0 | 200 MG
SDP
Catalog No. 5000339952
Encompass_Preferred
Sapitinib (AZD-8931) is a reversible, ATP competitive EGFR inhibitor with IC50s of 4, 3, and 4 nM for EGFR, ErbB2, and ErbB3 in cells, respectively. It demonstrates potent inhibitory effects on erbB2 in ligand-independent MCF-7 cl24 cells, significantly inhibits Akt phosphorylation, and induces apoptosis in human IBC cells.
  • Reversible, ATP competitive EGFR inhibitor
  • Potent inhibitory effect on erbB2 in ligand-independent MCF-7 cl24 cells
  • Significantly inhibits phosphorylation of Akt in SUM149 and FC-IBC-02 cells
  • Inhibits proliferation and induces apoptosis in human IBC cells
  • Active in xenograft tumor models responsive to EGFR inhibition alone or EGFR or erbB2 inhibition
  • Causes pharmacodynamic changes in proliferation and apoptosis markers in human tumor xenograft models
  • Favorable oral pharmacokinetics in rat and dog
  • Low human hepatocyte turnover
Catalog No. 50-003-39952 Supplier Medchemexpress LLC Supplier No. HY13050200MG
Sapitinib (AZD-8931) is a reversible, ATP competitive EGFR inhibitor with IC50s of 4, 3, and 4 nM for EGFR, ErbB2, and ErbB3 in cells, respectively. It demonstrates potent inhibitory effects on erbB2 in ligand-independent MCF-7 cl24 cells, significantly inhibits Akt phosphorylation, and induces apoptosis in human IBC cells.
  • Reversible, ATP competitive EGFR inhibitor
  • Potent inhibitory effect on erbB2 in ligand-independent MCF-7 cl24 cells
  • Significantly inhibits phosphorylation of Akt in SUM149 and FC-IBC-02 cells
  • Inhibits proliferation and induces apoptosis in human IBC cells
  • Active in xenograft tumor models responsive to EGFR inhibition alone or EGFR or erbB2 inhibition
  • Causes pharmacodynamic changes in proliferation and apoptosis markers in human tumor xenograft models
  • Favorable oral pharmacokinetics in rat and dog
  • Low human hepatocyte turnover