Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Saponin, Alfa Aesar

Manufacturer:  Alfa Aesar A1882022

Quantity 100g

 View more versions of this product

Catalog No. AAA1882022


  Check Availability
Add to cart

Specifications

Description

Applications
A glycoside used to permeabilize cellular membranesSaponin is used as dietary supplements and nutraceuticals and as adjuvants in the production of vaccines. It finds application in food, cosmetics and pharmaceutical sectors. It is used as a liquid soap, jewelry polish, detergent, pet shampoo, surfactant and wetting agent. It is also involved as anti-microbe to treat malaria, lower cholesterol, support to immune system, in bone strength and for hyper tension. Further, it is used as animal feed and an automobile cleaner.

Solubility
Soluble in cold water.

Notes
Incompatible with strong oxidizing agents.
Description & Specifications

Chemical Identifiers

8047-15-2
Powder
Saponin

Specifications

8047-15-2
Powder
100g
Saponin
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit