Learn More
Crescent Chemical Co Inc SAPONIN PURE
SDP
Supplier:  Crescent Chemical Co Inc SE34655.02
Encompass_Preferred
Saponin pure, DAB Isolated from Quillaja bark. For permeabilization of cells. Size - 250G Storage Conditions - +15 °C TO +30 °C Catalog Number - 34655.02 CAS 8047-15-2
Catalog No. 50-248-113