Learn More
Sigma Aldrich Fine Chemicals Biosciences Saporin from Saponaria off1MG
Catalog No. 502901756
Encompass
Saporin from Saponaria officinalis seeds has an N-terminal domain which is -stranded and a C-terminal domain which is helical. It is made up of 253 amino acids and has a molecular weight of 28 621Da.
Catalog No. 50-290-1756 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. S98961MG
Saporin from Saponaria officinalis seeds has an N-terminal domain which is -stranded and a C-terminal domain which is helical. It is made up of 253 amino acids and has a molecular weight of 28 621Da.