Learn More
IBL America sAPPβ-w (Mouse)
Supplier:  IBL America 27416
Encompass_Preferred
Solid phase sandwich ELISA for the determination of sAPP-w in mouse plasma and cell culture supernatant. For research use only, not for use in diagnostic procedures.As a primary antibody is coated on a plate, samples and standard are added into the wells for 1st reaction. After the reaction, HRP-conjugated secondary antibody is added into the wells for 2nd reaction. Afer washing away unbound the secondary antibody, Tetra Methyl Benzidine (TMB) is added to the wells and color develops.
Catalog No. 50-136-6568