Learn More
Meso Scale Discovery sAPPalpha/sAPPbeta Kit
SDP
Catalog No. 5000078827
Encompass
The SappAlpha/SappBeta Kit from Meso Scale Diagnostics measures alpha and beta secretase activity in biological samples It includes reagents plates substrates detection antibodies buffers and calibrators Using electrochemiluminescent detection it offers sensitive quantification of secretase activity The kit supports high-throughput screening of compounds affecting amyloid precursor protein processing Suitable for cell or tissue extracts it requires small sample volumes and includes reagents for 96 wells processing up to 40 samples in duplicate
Catalog No. 50-000-78827 Supplier Meso Scale Discovery Supplier No. K15120E2
Add to Cart
Add to Cart
Request a Quote
The SappAlpha/SappBeta Kit from Meso Scale Diagnostics measures alpha and beta secretase activity in biological samples It includes reagents plates substrates detection antibodies buffers and calibrators Using electrochemiluminescent detection it offers sensitive quantification of secretase activity The kit supports high-throughput screening of compounds affecting amyloid precursor protein processing Suitable for cell or tissue extracts it requires small sample volumes and includes reagents for 96 wells processing up to 40 samples in duplicate