Learn More
Medchemexpress LLC Sappanchalcone | 94344-54-4 | 99.5% | C16H14O5 | 5 MG
SDP
Catalog No. 5000357996
Encompass_Preferred
Sappanchalcone is a flavonoid isolated from *Caesalpinia sappan L.*, which induces caspase-dependent and AIF-dependent apoptosis in human colon cancer cells. It is intended for research use only.
  • Flavonoid isolated from *Caesalpinia sappan L.*
  • Induces caspase-dependent and AIF-dependent apoptosis
  • Shows statistically significant effect on HCT116 cells
  • Reduces MMP with significant increase in ROS levels
  • Triggers phosphorylation of p53
  • Inhibits LPS stimulated TNF alpha production in Balb/c mouse peritoneal macrophages
Catalog No. 50-003-57996 Supplier Medchemexpress LLC Supplier No. HY590015MG
Sappanchalcone is a flavonoid isolated from *Caesalpinia sappan L.*, which induces caspase-dependent and AIF-dependent apoptosis in human colon cancer cells. It is intended for research use only.
  • Flavonoid isolated from *Caesalpinia sappan L.*
  • Induces caspase-dependent and AIF-dependent apoptosis
  • Shows statistically significant effect on HCT116 cells
  • Reduces MMP with significant increase in ROS levels
  • Triggers phosphorylation of p53
  • Inhibits LPS stimulated TNF alpha production in Balb/c mouse peritoneal macrophages