Learn More
Cayman Chemical Sappanone A, Purity: ≥98%, CAS: 102067-84-5, 10 mg
SDP
Catalog No. 502987195
Encompass_Preferred
A homoisoflavonoid with diverse biological activities; inhibits FGFR1, KDR, c-Met, and c-Kit kinase activity (10 μM); inhibits viral NA (IC50s = 0.7, 1.1, and 1 μM for H1N1, H3N2, and H9N2 influenza viral NAs, respectively); has antibacterial and antifungal activities; reduces NO, IL-6, and PGE2 production in RAW264.7 cells; attenuates airway inflammation and mucus hypersecretion in a mouse model of ovalbumin-induced asthma
Catalog No. 50-298-7195 Supplier Cayman Chemical Supplier No. 2320510 MG
A homoisoflavonoid with diverse biological activities; inhibits FGFR1, KDR, c-Met, and c-Kit kinase activity (10 μM); inhibits viral NA (IC50s = 0.7, 1.1, and 1 μM for H1N1, H3N2, and H9N2 influenza viral NAs, respectively); has antibacterial and antifungal activities; reduces NO, IL-6, and PGE2 production in RAW264.7 cells; attenuates airway inflammation and mucus hypersecretion in a mouse model of ovalbumin-induced asthma