Learn More
Electron Microscopy Sciences Sapphire Burnishing Tool
SDP
Catalog No. 5028456
Encompass
This tool will out-perform any stone for the fine burnishing of soft or hard materials such as; miniature pivots, shafts, and pins that are made from hard steel, carbide and even ceramic. This precisely ground sapphire blade is 1.5 mm x 2.3 mm, 14 mm long (0.06x0.09x.56"). It is permanently mounted in a hexagonal gold anodized aluminum handle. Total length is 120 mm (4.75").
Catalog No. 50-284-56 Supplier Electron Microscopy Sciences Supplier No. 6216000
This tool will out-perform any stone for the fine burnishing of soft or hard materials such as; miniature pivots, shafts, and pins that are made from hard steel, carbide and even ceramic. This precisely ground sapphire blade is 1.5 mm x 2.3 mm, 14 mm long (0.06x0.09x.56"). It is permanently mounted in a hexagonal gold anodized aluminum handle. Total length is 120 mm (4.75").