Learn More
Glass Expansion Sapphire Capillary Injector with 2.0mm ID.A 2.0mm ID sapphire capillary injector suitable for use with aqueous matrices, including HF. Good for reduced boron background. Compatible with Thermo PRO Duo D-Torch (NC2046971).
SDP
Catalog No. 502703147
Encompass_Preferred
Sapphire Capillary Injector with 2.0mm ID.A 2.0mm ID sapphire capillary injector suitable for use with aqueous matrices, including HF. Good for reduced boron background. Compatible with Thermo PRO Duo D-Torch (NC2046971). When replacing your injector, it is recommended to replace the injector ferrule (31-808-2897), on average every 6 months.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3147 Supplier Glass Expansion Supplier No. 318082977
Sapphire Capillary Injector with 2.0mm ID.A 2.0mm ID sapphire capillary injector suitable for use with aqueous matrices, including HF. Good for reduced boron background. Compatible with Thermo PRO Duo D-Torch (NC2046971). When replacing your injector, it is recommended to replace the injector ferrule (31-808-2897), on average every 6 months.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001