Learn More
Azure Biosystems Sapphire FL Plate Holder
SDP
Catalog No. IS4103AZ Shop All Azure Biosystems Products
Change view
Click to view available options
Material:
Plastic
Catalog No. Material
IS4103AZ Plastic
1 options
Catalog No. IS4103AZ Supplier Azure Biosystems Supplier No. IS4103
Add to Cart
Add to Cart
Designed to hold up to six standard 96-well plates with optically clear bottoms for use with the Sapphire FL
  • Plate holder compatible with the Sapphire FL+
  • Not Autovclavable
  • Product intended to use with Sapphire FL+ Biomolecular Imager
  • Primary product material is made from Plastic
Specifications
Autoclavable Not Autoclavable
Type Plate Holder
Material Plastic
For Use With (Equipment) Sapphire FL+ Biomolecular Imager