Learn More
Glass Expansion Sapphire Injector 2.0mm
SDP
Supplier:  Glass Expansion 318082803
Encompass_Preferred
Sapphire Injector with 2.0mm ID.A 2.0mm ID sapphire injector (N0695495, 8003-0538, 50-190-7228, 31-808-2803) suitable for use with aqueous matrices, including HF. Good for reduced boron background. Compatible with PerkinElmer ICP-MS Elan (non-DRC), NexION 300/350 D-Torch (30-807-0504/30-808-2937) and standard torch. Suitable for torch adaptor 31-808-2825.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7152