Learn More
Glass Expansion Sapphire Injector 2.4mm for 7500 D-Torch
SDP
Catalog No. 501907213
Encompass_Preferred
Sapphire Capillary Injector with 2.4mm ID.A 2.4mm ID sapphire capillary injector suitable for use with aqueous matrices, including HF. Good for reduced boron background. Compatible with PerkinElmer1000/2000/5000 ICP-MS D-Torch (30-808-3927). Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7213 Supplier Glass Expansion Supplier No. 318083378
Sapphire Capillary Injector with 2.4mm ID.A 2.4mm ID sapphire capillary injector suitable for use with aqueous matrices, including HF. Good for reduced boron background. Compatible with PerkinElmer1000/2000/5000 ICP-MS D-Torch (30-808-3927). Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001