Learn More
Glass Expansion Sapphire Injector 2.4mm for D-Torch
SDP
Catalog No. 501907222
Encompass_Preferred
Sapphire Injector with 2.4mm ID.2.4mm ID sapphire injector (50-190-7222, 31-808-3414). This sapphire injector, with 2.4 mm I.D., is for use with Spectro EOP/SOP, Axial/Radial (Modula, CIROS, Genesis, ARCOS) ICP-OES, not for ARCOS II. Sapphire injectors are ideal for silicon and HF applications.Spectro EOP/SOP, Axial/Radial (Modula, CIROS, Genesis, ARCOS) ICP-OES Injectors - Compatible with Glass Expansion Axial D-Torch 50-190-7029 (30-808-3496) and Radial D-Torch 50-190-7023 (30-808-2928). Optional Tapered Quartz Injectors are 1.8 mm 50-190-7168 (31-808-2974), 2.5 mm 50-190-7252 (31-808-3723) and 3.0mm 31-808-3724, Tapered Alumina Injectors are 1.2mm 50-190-7191 (31-808-3238), 1.8mm 50-190-7166 (31-808-2963) and 2.4mm 50-190-7214 (31-808-3388). Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7222 Supplier Glass Expansion Supplier No. 318083414
Sapphire Injector with 2.4mm ID.2.4mm ID sapphire injector (50-190-7222, 31-808-3414). This sapphire injector, with 2.4 mm I.D., is for use with Spectro EOP/SOP, Axial/Radial (Modula, CIROS, Genesis, ARCOS) ICP-OES, not for ARCOS II. Sapphire injectors are ideal for silicon and HF applications.Spectro EOP/SOP, Axial/Radial (Modula, CIROS, Genesis, ARCOS) ICP-OES Injectors - Compatible with Glass Expansion Axial D-Torch 50-190-7029 (30-808-3496) and Radial D-Torch 50-190-7023 (30-808-2928). Optional Tapered Quartz Injectors are 1.8 mm 50-190-7168 (31-808-2974), 2.5 mm 50-190-7252 (31-808-3723) and 3.0mm 31-808-3724, Tapered Alumina Injectors are 1.2mm 50-190-7191 (31-808-3238), 1.8mm 50-190-7166 (31-808-2963) and 2.4mm 50-190-7214 (31-808-3388). Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001