Learn More
Glass Expansion Sapphire Capillary Injector with 2.0mm ID.A 2.0mm ID sapphire capillary injector suitable for use with aqueous matrices, including HF. Good for reduced boron background. Compatible with Agilent 700-ES Series Axial, Vista Axial D-Torch.
SDP
Catalog No. 502703137
Encompass_Preferred
Sapphire Capillary Injector with 2.0mm ID.A 2.0mm ID sapphire capillary injector suitable for use with aqueous matrices, including HF. Good for reduced boron background. Compatible with Agilent ICP-OES 700-ES Series Axial, Vista Axial D-Torch (501907021). When replacing your injector, it is recommended to replace the injector ferrule (31-808-2897), on average every 6 months.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3137 Supplier Glass Expansion Supplier No. 318083355
Sapphire Capillary Injector with 2.0mm ID.A 2.0mm ID sapphire capillary injector suitable for use with aqueous matrices, including HF. Good for reduced boron background. Compatible with Agilent ICP-OES 700-ES Series Axial, Vista Axial D-Torch (501907021). When replacing your injector, it is recommended to replace the injector ferrule (31-808-2897), on average every 6 months.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001