Learn More
Glass Expansion Sapphire Injector 2.4mm for D-Torch
SDP
Catalog No. 501907249
Encompass_Preferred
Sapphire Capillary Injector with 2.0mm ID.2.0mm ID Sapphire Capillary Injector. They are designed for organic solvents and non-HF matrices. Compatible with Spectro Arcos II EOP or MV Axial D-Torch (30-808-3657).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7249 Supplier Glass Expansion Supplier No. 318083718
Sapphire Capillary Injector with 2.0mm ID.2.0mm ID Sapphire Capillary Injector. They are designed for organic solvents and non-HF matrices. Compatible with Spectro Arcos II EOP or MV Axial D-Torch (30-808-3657).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001