Learn More
Glass Expansion Sapphire Injector 2.0mm
SDP
Catalog No. 501907171
Encompass_Preferred
Sapphire Injector with 2.0mm ID.2.0mm ID sapphire injector 50-190-7171 (31-808-3001). This sapphire injector, with 2.0mm ID, is for use with Shimadzu 7000/8000/9000/9800 Series ICP-OES and Thermo Element 1, Element 2, Neptune, X-Series ICP-MS. Sapphire injectors are ideal for silicon and HF applications.Compatible with Glass Expansion Semi Demountable Torch (SDT)-Shimadzu 7000/8000/9000/9800 Series ICP-OES –Quartz Body 31-808-1258, SDT 30-808-8039, and optional Tapered Alumina Injector 1.0mm is 31-808-0835. Thermo Element 1, Element 2, Neptune-Quartz Body 31-808-0451, SDT 30-808-0394, and SDT 30-808-0395. Optional Tapered Quartz Injector 2.0mm 50-190-7058 (31-808-0008), and Tapered Alumina Injector, 2.0mm 50-190-7066 (31-808-0027). Thermo X-Series ICP-MS-Quartz Body 31-808-1009. Optional Tapered Alumina Injectors 1.5mm 50-190-7125 (31-808-1013); Tapered Quartz Injector 1.5mm 50-190-7126 (31-808-1014).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7171 Supplier Glass Expansion Supplier No. 318083001
Sapphire Injector with 2.0mm ID.2.0mm ID sapphire injector 50-190-7171 (31-808-3001). This sapphire injector, with 2.0mm ID, is for use with Shimadzu 7000/8000/9000/9800 Series ICP-OES and Thermo Element 1, Element 2, Neptune, X-Series ICP-MS. Sapphire injectors are ideal for silicon and HF applications.Compatible with Glass Expansion Semi Demountable Torch (SDT)-Shimadzu 7000/8000/9000/9800 Series ICP-OES –Quartz Body 31-808-1258, SDT 30-808-8039, and optional Tapered Alumina Injector 1.0mm is 31-808-0835. Thermo Element 1, Element 2, Neptune-Quartz Body 31-808-0451, SDT 30-808-0394, and SDT 30-808-0395. Optional Tapered Quartz Injector 2.0mm 50-190-7058 (31-808-0008), and Tapered Alumina Injector, 2.0mm 50-190-7066 (31-808-0027). Thermo X-Series ICP-MS-Quartz Body 31-808-1009. Optional Tapered Alumina Injectors 1.5mm 50-190-7125 (31-808-1013); Tapered Quartz Injector 1.5mm 50-190-7126 (31-808-1014).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001