Learn More
Perkin Elmer US LLC Sapphire Injector for Demountable ZipTorch, 1.5 mm I.D. Optima 8x00
Catalog No. 502112267
Encompass_Preferred
This Demountable Injector is for use with Optima 8x00 ICP-OES series instruments. It is designed for use with the quartz, organics and silicon nitride demountable ZipTorches. The demountable design allows the injector to be removed and replaced efficiently for cleaning without dismounting the torch.
Catalog No. 50-211-2267 Supplier Perkin Elmer US LLC Supplier No. N0782004
This Demountable Injector is for use with Optima 8x00 ICP-OES series instruments. It is designed for use with the quartz, organics and silicon nitride demountable ZipTorches. The demountable design allows the injector to be removed and replaced efficiently for cleaning without dismounting the torch.