Learn More
Glass Expansion Sapphire Capillary Injector with 2.0mm ID. 2.0mm ID capillary sapphire injector. For use with Avio 200/220 and 500/550 ICP-OES series. Sapphire Injectors are recommended for silicon, environmental, geochemistry & various HF applications.
SDP
Catalog No. 502703115
Encompass_Preferred
Sapphire Capillary Injector with 2.0mm ID. 2.0mm ID capillary sapphire injector (N0791184). For use with Avio 200/220 and 500/550 ICP-OES series. Sapphire Injectors are recommended for silicon, environmental, geochemistry & various HF applications. Compatible with PerkinElmer Avio 200/220 D-Torch (P/N 50-190-7038), Avio 500/550 D-Torch (P/N 50-190-7039), Avio 200/220 Adjustable torch body (P/N 50-211-3618) and Avio 500/550 Adjustable torch body (P/N 50-211-2626).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3115 Supplier Glass Expansion Supplier No. 318083784
Sapphire Capillary Injector with 2.0mm ID. 2.0mm ID capillary sapphire injector (N0791184). For use with Avio 200/220 and 500/550 ICP-OES series. Sapphire Injectors are recommended for silicon, environmental, geochemistry & various HF applications. Compatible with PerkinElmer Avio 200/220 D-Torch (P/N 50-190-7038), Avio 500/550 D-Torch (P/N 50-190-7039), Avio 200/220 Adjustable torch body (P/N 50-211-3618) and Avio 500/550 Adjustable torch body (P/N 50-211-2626).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001