Learn More
Electron Microscopy Sciences Sapphire Blade 1.5" x 0.4" x .03
SDP
Catalog No. 5030472
Encompass_Preferred
The Sapphire Knife offers smooth, consistent and thinner sections than are usually obtainable with stainless steel blades. The high-precision knife is for use with all Vibratomes and Tissue Sectioners, as well as our Oscillating Tissue Slicers. The blade allows for sectioning as thin as 10 microns. Offering many years of cutting surface, the durability of far outlasts other comparable blades on the market. Made of synthetic Single Crystal Sapphire.
Size: 1.5" x 0.4" x .03
Catalog No. 50-304-72 Supplier Electron Microscopy Sciences Supplier No. 93060
The Sapphire Knife offers smooth, consistent and thinner sections than are usually obtainable with stainless steel blades. The high-precision knife is for use with all Vibratomes and Tissue Sectioners, as well as our Oscillating Tissue Slicers. The blade allows for sectioning as thin as 10 microns. Offering many years of cutting surface, the durability of far outlasts other comparable blades on the market. Made of synthetic Single Crystal Sapphire.
Size: 1.5" x 0.4" x .03