Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One Sapphire Polypropylene Full-Skirted 384-well PCR Microplates

Catalog No. 07000044 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07-000-044 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-000-044 Supplier Greiner Bio-One Supplier No. 785290

missing translation for 'validMainframeMsg'

Suitable for adhesive sealers and heat sealing

Blue alphanumeric well coding
TRUSTED_SUSTAINABILITY

Specifications

Breakapart No
Lid Without Lid
Array 16 x 24
Autoclavable Not Autoclavable
Binding Type Non Binding
Color Clear
Material Polypropylene
No. of Wells 384
Volume (Metric) Working 25 μL
Sterility Non-sterile
Surface Treatment Untreated
Well Shape Conical Bottom (V-well)
Dnase Free Yes
For Use With (Application) PCR
Max. RCF 4800 x g (swinging bucket rotor)
No. per Case 60
No. per Pack 15
Plate Compatibility Suitable for ABI
RNase Free Yes
Well Color Clear
Dimensions (L x W x H) 127.76 x 85.48 x 9.6 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.