Learn More
Greiner Bio-One Sapphire Polypropylene Full-Skirted 384-well PCR Microplates
Catalog No. 07000045 Shop All Greiner Bio-One Products
Catalog No. 07-000-045 Supplier Greiner Bio-One Supplier No. 785201
Suitable for adhesive sealers and heat sealing
Blue alphanumeric well coding
TRUSTED_SUSTAINABILITY
Specifications
Array 16 x 24
Autoclavable Not Autoclavable
Material Polypropylene
No. of Wells 384
For Use With (Application) PCR