Learn More
TA Instruments Sapphire Specific Heat Material DSC Hermetic Sample Pans, Heat Capacity Calibration, One Sapphire Disc (0.135 Inch OD X 0. 020 Inch Height), DSC Hermetic Sample Pans
Catalog No. 5000580910 Shop All TA Instruments Products
Encompass_Preferred
Waters TA Instruments Sapphire Specific Heat Material for DSC Hermetic Sample Pans is used for heat capacity calibration of the DSC cell. Includes one sapphire disc (0.135 inch OD x 0.020 inch height) for use in hermetic DSC sample pans.
Catalog No. 50-005-80910 Supplier TA Instruments Supplier No. 915079.902
Waters TA Instruments Sapphire Specific Heat Material for DSC Hermetic Sample Pans is used for heat capacity calibration of the DSC cell. Includes one sapphire disc (0.135 inch OD x 0.020 inch height) for use in hermetic DSC sample pans.