Learn More
TA Instruments Sapphire Specific Heat Material DSC Standard Sample Pans, Heat Capacity Calibration, One Sapphire Disc (0.16 Inch OD X 0. 020 Inch Height), Standard DSC Sample Pans
Catalog No. 5000580609 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Sapphire Specific Heat Material for DSC Standard Sample Pans includes one sapphire disc (0.16 inch OD x 0.020 inch height) used for heat capacity calibration of the DSC cell in standard pans.
Catalog No. 50-005-80609 Supplier TA Instruments Supplier No. 915079.901
Waters TA Instruments Sapphire Specific Heat Material for DSC Standard Sample Pans includes one sapphire disc (0.16 inch OD x 0.020 inch height) used for heat capacity calibration of the DSC cell in standard pans.