Learn More
TA Instruments Sapphire Specific Heat Material DSC Standard Sample Pans, Heat Capacity Calibration, One Sapphire Disc (0.187 Inch OD X 0. 034 Inch Height), Standard DSC Sample Pans
Catalog No. 5000580886 Shop All TA Instruments Products
Encompass_Preferred
Waters TA Instruments Sapphire Specific Heat Material for DSC Standard Sample Pans includes one sapphire disc (0.187 inch OD x 0.034 inch height) used for heat capacity calibration of the DSC cell in standard pans.
Catalog No. 50-005-80886 Supplier TA Instruments Supplier No. 915079.903
Waters TA Instruments Sapphire Specific Heat Material for DSC Standard Sample Pans includes one sapphire disc (0.187 inch OD x 0.034 inch height) used for heat capacity calibration of the DSC cell in standard pans.