Learn More
Ted Pella Inc SAPPHIRE SUBSTRATE DISC
SDP
Catalog No. NC0954128
Encompass
Sapphire Substrate Disc, �25.4mm x 0.5mm (�1" x 0.020"), polished. The PELCO� Sapphire Discs or wafers are made from high purity clear sapphire (Al2O3), ground and optically clear polished on both sides. (Technical Data for Sapphire). Sapphire has a wide transmission range; from 150 to 6000nm with low absorbance in the 300 to 4500nm range. The discs are ideal as substrates for thin film research and are suitable for optical research. Excellent resistance against a wide range of chemicals. Surface polish to one microinch or better on each face. Tolerances are �0.001" (0.025mm) in diameter and thickness; flatness to 0.0003" (0.0008mm) or better.
Catalog No. NC0954128 Supplier Ted Pella Inc Supplier No. 160051020
Sapphire Substrate Disc, �25.4mm x 0.5mm (�1" x 0.020"), polished. The PELCO� Sapphire Discs or wafers are made from high purity clear sapphire (Al2O3), ground and optically clear polished on both sides. (Technical Data for Sapphire). Sapphire has a wide transmission range; from 150 to 6000nm with low absorbance in the 300 to 4500nm range. The discs are ideal as substrates for thin film research and are suitable for optical research. Excellent resistance against a wide range of chemicals. Surface polish to one microinch or better on each face. Tolerances are �0.001" (0.025mm) in diameter and thickness; flatness to 0.0003" (0.0008mm) or better.