Learn More
Takara Bio SapphireAmp® Fast PCR Master Mix, 800 Rxns
SDP
Catalog No. 50444018
Encompass
A hot-start 2X PCR master mix with dye. SapphireAmp Fast PCR mix is well-suited for E.coli-based colony PCR, and colony checks can be completed in about 1 hour. Reactions performed with this mix can be loaded directly onto a gel for electrophoresis. Restriction digestion of PCR products is possible in SapphireAmp reaction buffer. 800 Rxns
Catalog No. 50-444-018 Supplier Takara Bio Supplier No. RR350B
A hot-start 2X PCR master mix with dye. SapphireAmp Fast PCR mix is well-suited for E.coli-based colony PCR, and colony checks can be completed in about 1 hour. Reactions performed with this mix can be loaded directly onto a gel for electrophoresis. Restriction digestion of PCR products is possible in SapphireAmp reaction buffer. 800 Rxns